accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C(S)(S)NHOHCH3OOH3CCH3H3C

CAS RN 106946-74-1

IUPAC Name: tert-butyl N-[(2S,3S)-1-hydroxy-3-methylpentan-2-yl]carbamate
Synonyms: tert-butyl N-[(2S,3S)-1-hydroxy-3-methylpentan-2-yl]carbamate
Molecular Weight (g/mol): 217.31
Molecular Formula: C11H23NO3
InChi Key: BPLDQMXXYMKQPW-DTWKUNHWSA-N
SMILES: CC[C@H](C)[C@@H](CO)NC(=O)OC(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results