accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 107-03-9

IUPAC Name: propane-1-thiol
Synonyms: propanethiol n-thiopropyl alcohol sodium salt propanethiol thiopropyl alcohol
Molecular Weight (g/mol): 76.16
Molecular Formula: C3H8S
InChi Key: SUVIGLJNEAMWEG-UHFFFAOYSA-N
SMILES: CCCS
Molecular Weight (g/mol) 
  • (5)
  • (2)
Boiling Point 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (5)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)
  • (4)
  • (1)