accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 107-04-0

IUPAC Name: 1-bromo-2-chloroethane
Synonyms: 1-bromo-2-chloroethane sym-chlorobromoethane caswell no. 436B ethylene chlorobromide epa pesticide chemical code 042001 s-chlorobromoethane C2-H4-br-cl ccris 816 1,2-chlorobromoethane 1,2-bromochloroethane
Molecular Weight (g/mol): 143.41
Molecular Formula: C2H4BrCl
InChi Key: IBYHHJPAARCAIE-UHFFFAOYSA-N
SMILES: ClCCBr
Boiling Point 
  • (3)
  • (3)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (6)
  • (4)
Percent Purity 
  • (3)
  • (7)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (3)
  • (5)
  • (3)
  • (3)
  • (2)
  • (2)
  • (6)
  • (4)
  • (3)
  • (7)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (5)