accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHCl

CAS RN 107-07-3

IUPAC Name: 2-chloroethan-1-ol
Synonyms: 2-chloroethanol chloroethylowy alkohol ethylchlorohydrin chlorohydrin ethylene glycol ethylene chlorohydrin (substance) aethylenechlorhydrin ethylene chlorhydrin glycol chlorohydrin 2-cloroetanolo ethene, chlorohydrin
Molecular Weight (g/mol): 80.51
Molecular Formula: C2H5ClO
InChi Key: SZIFAVKTNFCBPC-UHFFFAOYSA-N
SMILES: OCCCl
Molecular Weight (g/mol) 
  • (3)
  • (4)
Boiling Point 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (3)
  • (4)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)