accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHN

CAS RN 107-16-4

IUPAC Name: 2-hydroxyacetonitrile
Synonyms: glycolonitrile
Molecular Weight (g/mol): 57.05
Molecular Formula: C2H3NO
InChi Key: LTYRAPJYLUPLCI-UHFFFAOYSA-N
SMILES: OCC#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results