accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHH2C

CAS RN 107-18-6

IUPAC Name: prop-2-en-1-ol
Synonyms: allyl alcohol caswell no. 026 rcra waste number P005 epa pesticide chemical code 068401 alcool allilco weed drench allylalkohol allilowy alkohol alcool allylique propen-1-ol-3
Molecular Weight (g/mol): 58.08
Molecular Formula: C3H6O
InChi Key: XXROGKLTLUQVRX-UHFFFAOYSA-N
SMILES: OCC=C
Molecular Weight (g/mol) 
  • (9)
  • (3)
Boiling Point 
  • (3)
  • (3)
  • (2)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (5)
  • (1)
  • (6)
  • (9)
  • (3)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (6)
  • (3)
  • (1)