accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClO

CAS RN 107-20-0

IUPAC Name: 2-chloroacetaldehyde
Synonyms: chloroacetaldehyde rcra waste number P023 chloroacetaldehyde (substance) ccris 856 chloroaldehyde chloroacetaldehyde monomer
Molecular Weight (g/mol): 78.5
Molecular Formula: C2H3ClO
InChi Key: QSKPIOLLBIHNAC-UHFFFAOYSA-N
SMILES: ClCC=O
Molecular Weight (g/mol) 
  • (1)
  • (1)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (5)
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)