CAS RN 107-41-5
2-Methyl-2,4-pentanediol, 99+%
CAS: 107-41-5 Molecular Formula: C6H14O2 Molecular Weight (g/mol): 118.18 InChI Key: SVTBMSDMJJWYQN-UHFFFAOYSA-N Synonym: hexylene glycol,2-methyl-2,4-pentanediol,pinakon,2,4-dihydroxy-2-methylpentane,2,4-pentanediol, 2-methyl,diolane,isol,4-methyl-2,4-pentanediol,1,1,3-trimethyltrimethylenediol,2-methyl pentane-2,4-diol PubChem CID: 7870 ChEBI: CHEBI:62995 IUPAC Name: 2-methylpentane-2,4-diol SMILES: CC(CC(C)(C)O)O
(+/-)-2-Methyl-2,4-pentanediol, 98%
CAS: 107-41-5 Molecular Formula: C6H14O2 Molecular Weight (g/mol): 118.176 MDL Number: MFCD00004547 InChI Key: SVTBMSDMJJWYQN-UHFFFAOYSA-N Synonym: hexylene glycol,2-methyl-2,4-pentanediol,pinakon,2,4-dihydroxy-2-methylpentane,2,4-pentanediol, 2-methyl,diolane,isol,4-methyl-2,4-pentanediol,1,1,3-trimethyltrimethylenediol,2-methyl pentane-2,4-diol PubChem CID: 7870 ChEBI: CHEBI:62995 IUPAC Name: 2-methylpentane-2,4-diol SMILES: CC(CC(C)(C)O)O
Hexylene Glycol, Reagent, 98%, Spectrum™ Chemical
CAS: 107-41-5 Molecular Formula: C6H14O2 Molecular Weight (g/mol): 118.18 InChI Key: SVTBMSDMJJWYQN-UHFFFAOYNA-N IUPAC Name: 2-methylpentane-2,4-diol SMILES: CC(O)CC(C)(C)O
2-Methyl-2,4-pentanediol, 99%
CAS: 107-41-5 Molecular Formula: C6H14O2 Molecular Weight (g/mol): 118.18 MDL Number: MFCD00004547 InChI Key: SVTBMSDMJJWYQN-UHFFFAOYSA-N Synonym: hexylene glycol,2-methyl-2,4-pentanediol,pinakon,2,4-dihydroxy-2-methylpentane,2,4-pentanediol, 2-methyl,diolane,isol,4-methyl-2,4-pentanediol,1,1,3-trimethyltrimethylenediol,2-methyl pentane-2,4-diol PubChem CID: 7870 ChEBI: CHEBI:62995 IUPAC Name: 2-methylpentane-2,4-diol SMILES: CC(CC(C)(C)O)O
2-Methylpentane-2,4-diol 99.0+%, TCI America™
CAS: 107-41-5 Molecular Formula: C6H14O2 Molecular Weight (g/mol): 118.176 MDL Number: MFCD00004547 InChI Key: SVTBMSDMJJWYQN-UHFFFAOYSA-N Synonym: hexylene glycol,2-methyl-2,4-pentanediol,pinakon,2,4-dihydroxy-2-methylpentane,2,4-pentanediol, 2-methyl,diolane,isol,4-methyl-2,4-pentanediol,1,1,3-trimethyltrimethylenediol,2-methyl pentane-2,4-diol PubChem CID: 7870 ChEBI: CHEBI:62995 IUPAC Name: 2-methylpentane-2,4-diol SMILES: CC(CC(C)(C)O)O
Sigma Aldrich Hexylene glycol
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Hexylene glycol
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
2-Methyl-2,4-Pentanediol, SPEX CertiPrep™
CAS: 107-41-5 Molecular Formula: C6H14O2 Molecular Weight (g/mol): 118.176 InChI Key: SVTBMSDMJJWYQN-UHFFFAOYSA-N PubChem CID: 7870 ChEBI: CHEBI:62995 IUPAC Name: 2-methylpentane-2,4-diol SMILES: CC(CC(C)(C)O)O