accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 107-45-9

IUPAC Name: 2,4,4-trimethylpentan-2-amine
Synonyms: 2,4,4-trimethylpentan-2-amine
Molecular Weight (g/mol): 129.25
Molecular Formula: C8H19N
InChi Key: QIJIUJYANDSEKG-UHFFFAOYSA-N
SMILES: CC(C)(C)CC(C)(C)N
Boiling Point 
  • (2)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (4)