accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 107-47-1

H3CCH3H3CSCH3CH3H3C

CAS RN 107-47-1

IUPAC Name: 2-(tert-butylsulfanyl)-2-methylpropane
Synonyms: 2-(tert-butylsulfanyl)-2-methylpropane
Molecular Weight (g/mol): 146.29
Molecular Formula: C8H18S
InChi Key: LNMBCRKRCIMQLW-UHFFFAOYSA-N
SMILES: CC(C)(C)SC(C)(C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results