accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OPOOCH3OPOOCH3OCH3

CAS RN 107-49-3

IUPAC Name: diethyl [(diethoxyphosphoryl)oxy]phosphonate
Synonyms: tetraethyl pyrophosphate
Molecular Weight (g/mol): 290.19
Molecular Formula: C8H20O7P2
InChi Key: IDCBOTIENDVCBQ-UHFFFAOYSA-N
SMILES: CCOP(=O)(OCC)OP(=O)(OCC)OCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results