accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 107-58-4

IUPAC Name: N-tert-butylprop-2-enamide
Synonyms: tert-butylacrylamide
Molecular Weight (g/mol): 127.19
Molecular Formula: C7H13NO
InChi Key: XFHJDMUEHUHAJW-UHFFFAOYSA-N
SMILES: CC(C)(C)NC(=O)C=C
Molecular Weight (g/mol) 
  • (1)
  • (6)
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (4)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (4)
  • (1)
  • (6)
  • (2)
  • (5)
  • (4)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)