accessibility menu, dialog, popup

UserName

Viewing profile as user:
OSOO

CAS RN 107-61-9

IUPAC Name: 1,4λ⁶-oxathiane-4,4-dione
Synonyms: 1,4-oxathiane, 4,4-dioxide
Molecular Weight (g/mol): 136.17
Molecular Formula: C4H8O3S
InChi Key: WWRUZECKUVNAPB-UHFFFAOYSA-N
SMILES: O=S1(=O)CCOCC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results