accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOCH3CH3H3C

CAS RN 107-71-1

IUPAC Name: tert-butyl ethaneperoxoate
Synonyms: tert-butyl ethaneperoxoate
Molecular Weight (g/mol): 132.16
Molecular Formula: C6H12O3
InChi Key: SWAXTRYEYUTSAP-UHFFFAOYSA-N
SMILES: CC(=O)OOC(C)(C)C
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)