accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 107-87-9

IUPAC Name: pentan-2-one
Synonyms: 2-pentanone fema number 2842 metylopropyloketon 2-pentanone (natural) methyl-propyl-cetone
Molecular Weight (g/mol): 86.13
Molecular Formula: C5H10O
InChi Key: XNLICIUVMPYHGG-UHFFFAOYSA-N
SMILES: CCCC(C)=O
Molecular Weight (g/mol) 
  • (13)
  • (6)
Boiling Point 
  • (3)
  • (6)
  • (2)
  • (7)
Percent Purity 
  • (3)
  • (2)
  • (7)
  • (4)
  • (2)
Quantity 
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (13)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (10)
  • (7)
  • (1)
  • (2)
  • (9)
  • (13)
  • (6)
  • (3)
  • (6)
  • (2)
  • (7)
  • (3)
  • (2)
  • (7)
  • (4)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (13)
  • (4)
  • (7)
  • (1)