CAS RN 107-96-0
3-Mercaptopropionic acid, 99%
CAS: 107-96-0 Molecular Formula: C3H6O2S Molecular Weight (g/mol): 106.139 MDL Number: MFCD00004897 InChI Key: DKIDEFUBRARXTE-UHFFFAOYSA-N Synonym: 3-mercaptopropionic acid,3-mercaptopropanoic acid,3-thiopropionic acid,3-thiopropanoic acid,propanoic acid, 3-mercapto,beta-mercaptopropionic acid,3mpa,mercaptopropionic acid,beta-thiopropionic acid,2-mercaptoethanecarboxylic acid PubChem CID: 6514 ChEBI: CHEBI:44111 IUPAC Name: 3-sulfanylpropanoic acid SMILES: C(CS)C(=O)O
3-Mercaptopropionic Acid 98.0+%, TCI America™
CAS: 107-96-0 Molecular Formula: C3H6O2S Molecular Weight (g/mol): 106.139 MDL Number: MFCD00004897 InChI Key: DKIDEFUBRARXTE-UHFFFAOYSA-N Synonym: 3-mercaptopropionic acid,3-mercaptopropanoic acid,3-thiopropionic acid,3-thiopropanoic acid,propanoic acid, 3-mercapto,beta-mercaptopropionic acid,3mpa,mercaptopropionic acid,beta-thiopropionic acid,2-mercaptoethanecarboxylic acid PubChem CID: 6514 ChEBI: CHEBI:44111 IUPAC Name: 3-sulfanylpropanoic acid SMILES: C(CS)C(=O)O
3-Mercaptopropionic acid, 98%
CAS: 107-96-0 Molecular Formula: C3H6O2S Molecular Weight (g/mol): 106.14 MDL Number: MFCD00004897 InChI Key: DKIDEFUBRARXTE-UHFFFAOYSA-N Synonym: 3-mercaptopropionic acid,3-mercaptopropanoic acid,3-thiopropionic acid,3-thiopropanoic acid,propanoic acid, 3-mercapto,beta-mercaptopropionic acid,3mpa,mercaptopropionic acid,beta-thiopropionic acid,2-mercaptoethanecarboxylic acid PubChem CID: 6514 ChEBI: CHEBI:44111 IUPAC Name: 3-sulfanylpropanoic acid SMILES: C(CS)C(=O)O
Sigma Aldrich 3-Mercaptopropionic acid
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
MP Biomedicals, Inc βMercaptopropionic Acid, MP Biomedicals
CAS: 107-96-0 Molecular Formula: C3H6O2S Molecular Weight (g/mol): 106.139 InChI Key: DKIDEFUBRARXTE-UHFFFAOYSA-N Synonym: 3-mercaptopropionic acid,3-mercaptopropanoic acid,3-thiopropionic acid,3-thiopropanoic acid,propanoic acid, 3-mercapto,beta-mercaptopropionic acid,3mpa,mercaptopropionic acid,beta-thiopropionic acid,2-mercaptoethanecarboxylic acid PubChem CID: 6514 ChEBI: CHEBI:44111 IUPAC Name: 3-sulfanylpropanoic acid SMILES: C(CS)C(=O)O