accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 107-97-1

IUPAC Name: 2-(methylamino)acetic acid
Synonyms: sarcosine MeGly sarcosylate, magnesium Sar magnesium sarcosylate methylglycine
Molecular Weight (g/mol): 89.09
Molecular Formula: C3H7NO2
InChi Key: FSYKKLYZXJSNPZ-UHFFFAOYSA-N
SMILES: CNCC(O)=O
Percent Purity 
  • (3)
  • (3)
  • (3)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results