accessibility menu, dialog, popup

UserName

Viewing profile as user:
HNOOOHOHHOHOClBr

CAS RN 107021-38-5

IUPAC Name: 2-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol
Synonyms: 2-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular Weight (g/mol): 408.63
Molecular Formula: C14H15BrClNO6
InChi Key: OPIFSICVWOWJMJ-UHFFFAOYNA-N
SMILES: OCC1OC(OC2=CNC3=CC=C(Br)C(Cl)=C23)C(O)C(O)C1O
Physical Form 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Grade 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)