accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3H3CH3CPOH3COH3CCH3CH3(?)

CAS RN 1070663-78-3

IUPAC Name: dicyclohexyl[3,6-dimethoxy-2',4',6'-tris(propan-2-yl)-[1,1'-biphenyl]-2-yl]phosphane
Synonyms: dicyclohexyl({2',4',6'-triisopropyl-3,6-dimethoxy-[1,1'-biphenyl]-2-yl})phosphane
Molecular Weight (g/mol): 536.78
Molecular Formula: C35H53O2P
InChi Key: WDVGNXKCFBOKDF-UHFFFAOYSA-N
SMILES: COC1=C(P(C2CCCCC2)C2CCCCC2)C(=C(OC)C=C1)C1=C(C=C(C=C1C(C)C)C(C)C)C(C)C
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)