accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClSiClClN

CAS RN 1071-22-3

IUPAC Name: 3-(trichlorosilyl)propanenitrile
Synonyms: 3-trichlorosilylpropionitrile
Molecular Weight (g/mol): 188.51
Molecular Formula: C3H4Cl3NSi
InChi Key: OLBGECWYBGXCNV-UHFFFAOYSA-N
SMILES: Cl[Si](Cl)(Cl)CCC#N
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results