accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClSiClClN

CAS RN 1071-27-8

IUPAC Name: 4-(trichlorosilyl)butanenitrile
Synonyms: 4-(trichlorosilyl)butanenitrile
Molecular Weight (g/mol): 202.53
Molecular Formula: C4H6Cl3NSi
InChi Key: HMFFOEBLYHLRQN-UHFFFAOYSA-N
SMILES: Cl[Si](Cl)(Cl)CCCC#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results