accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1071-73-4

IUPAC Name: 5-hydroxypentan-2-one
Synonyms: 5-hydroxy-2-pentanone
Molecular Weight (g/mol): 102.13
Molecular Formula: C5H10O2
InChi Key: JSHPTIGHEWEXRW-UHFFFAOYSA-N
SMILES: CC(=O)CCCO
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)