CAS RN 1071224-34-4
IUPAC Name:
7-bromo-2,1,3-benzothiadiazole-4-carbaldehyde
Synonyms:
7-bromo-2,1,3-benzothiadiazole-4-carbaldehyde
Molecular Weight (g/mol):
243.08
Molecular Formula:
C7H3BrN2OS
InChi Key:
VPDPNJVAWKCZEH-UHFFFAOYSA-N
SMILES:
BrC1=CC=C(C=O)C2=NSN=C12
1
–
2
of
2
results
1
7-Bromo-2,1,3-benzothiadiazole-4-carboxaldehyde 98.0+%, TCI America™
CAS: 1071224-34-4 Molecular Formula: C7H3BrN2OS Molecular Weight (g/mol): 243.078 MDL Number: MFCD23704428 InChI Key: VPDPNJVAWKCZEH-UHFFFAOYSA-N Synonym: 7-Bromo-4-formyl-2,1,3-benzothiadiazole PubChem CID: 58497962 IUPAC Name: 4-bromo-2,1,3-benzothiadiazole-7-carbaldehyde SMILES: C1=C(C2=NSN=C2C(=C1)Br)C=O
2
Sigma Aldrich 7-Bromo-2,1,3-benzothiadiazole-4-carboxaldehyde
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.