accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOH3CCH3OOH3CCH2CH2

CAS RN 1071466-61-9

IUPAC Name: 4-{2-[4-(acetyloxy)-3-(prop-2-en-1-yl)phenyl]propan-2-yl}-2-(prop-2-en-1-yl)phenyl acetate
Synonyms: 4-{2-[4-(acetyloxy)-3-(prop-2-en-1-yl)phenyl]propan-2-yl}-2-(prop-2-en-1-yl)phenyl acetate
Molecular Weight (g/mol): 392.5
Molecular Formula: C25H28O4
InChi Key: GJTZQXNRNNDGRG-UHFFFAOYSA-N
SMILES: CC(=O)OC1=C(CC=C)C=C(C=C1)C(C)(C)C1=CC(CC=C)=C(OC(C)=O)C=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)