accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1072-71-5

IUPAC Name: 1,3,4-thiadiazolidine-2,5-dithione
Synonyms: 1,3,4-thiadiazolidine-2,5-dithione
Molecular Weight (g/mol): 150.23
Molecular Formula: C2H2N2S3
InChi Key: BIGYLAKFCGVRAN-UHFFFAOYSA-N
SMILES: S=C1NNC(=S)S1
Molecular Weight (g/mol) 
  • (9)
  • (1)
Percent Purity 
  • (2)
  • (2)
  • (6)
Quantity 
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
Physical Form 
  • (4)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (4)
  • (5)
  • (9)
  • (1)
  • (2)
  • (2)
  • (6)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (4)
  • (3)