accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONH

CAS RN 1072-82-8

IUPAC Name: 1-(1H-pyrrol-3-yl)ethan-1-one
Synonyms: 1-(1H-pyrrol-3-yl)ethanone
Molecular Weight (g/mol): 109.13
Molecular Formula: C6H7NO
InChi Key: KHHSXHXUQVNBGA-UHFFFAOYSA-N
SMILES: CC(=O)C1=CNC=C1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results