accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1072-84-0

IUPAC Name: 1H-imidazole-5-carboxylic acid
Synonyms: imidazole-4-carboxylic acid
Molecular Weight (g/mol): 112.09
Molecular Formula: C4H4N2O2
InChi Key: NKWCGTOZTHZDHB-UHFFFAOYSA-N
SMILES: OC(=O)C1=CN=CN1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (3)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)