accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1072-85-1

IUPAC Name: 1-bromo-2-fluorobenzene
Synonyms: 1-bromo-2-fluorobenzene
Molecular Weight (g/mol): 175
Molecular Formula: C6H4BrF
InChi Key: IPWBFGUBXWMIPR-UHFFFAOYSA-N
SMILES: FC1=CC=CC=C1Br
Percent Purity 
  • (3)
  • (7)
Quantity 
  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (3)
  • (5)
  • (3)
  • (7)
  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (5)