accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1072944-23-0

IUPAC Name: (6-bromo-2,4-dimethylpyridin-3-yl)boronic acid
Synonyms: 6-bromo-2,4-dimethylpyridin-3-ylboronic acid
Molecular Weight (g/mol): 229.87
Molecular Formula: C7H9BBrNO2
InChi Key: GXXITYQLWOMIEM-UHFFFAOYSA-N
SMILES: CC1=CC(Br)=NC(C)=C1B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results