accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1073-06-9

IUPAC Name: 1-bromo-3-fluorobenzene
Synonyms: M-bromofluorobenzene
Molecular Weight (g/mol): 175
Molecular Formula: C6H4BrF
InChi Key: QDFKKJYEIFBEFC-UHFFFAOYSA-N
SMILES: FC1=CC=CC(Br)=C1
Boiling Point 
  • (2)
  • (4)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
  • (6)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (2)
  • (6)
  • (2)
  • (4)
  • (1)
  • (2)
  • (2)
  • (1)
  • (6)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (4)