accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1073-21-8

IUPAC Name: 2,4-dimethylpyridin-3-amine
Synonyms: 2,4-dimethylpyridin-3-amine
Molecular Weight (g/mol): 122.17
Molecular Formula: C7H10N2
InChi Key: FSYHJEBPLDNXAM-UHFFFAOYSA-N
SMILES: CC1=C(N)C(C)=NC=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results