accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1073353-89-5

IUPAC Name: 2-(2-fluoro-4-nitrophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-(2-fluoro-4-nitrophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 267.06
Molecular Formula: C12H15BFNO4
InChi Key: QYXHQOSFZITWSU-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=C(F)C=C(C=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results