accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1073354-07-0

IUPAC Name: 2-[2-(1,3-dioxolan-2-yl)ethyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-[2-(1,3-dioxolan-2-yl)ethyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 228.1
Molecular Formula: C11H21BO4
InChi Key: DNBRLKJRBDIKOO-UHFFFAOYSA-N
SMILES: CC1(C)OB(CCC2OCCO2)OC1(C)C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results