accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOOH3CCH3H3CH3CNNH2

CAS RN 1073354-97-8

IUPAC Name: 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine
Synonyms: 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine
Molecular Weight (g/mol): 220.08
Molecular Formula: C11H17BN2O2
InChi Key: FSJAKASJCJZKET-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=C(N)N=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results