accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1073371-77-3

IUPAC Name: 4-chloro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
Synonyms: 4-chloro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
Molecular Weight (g/mol): 253.53
Molecular Formula: C12H17BClNO2
InChi Key: GVJZHGCVSYBPIM-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=C(N)C=CC(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results