accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1073372-02-7

IUPAC Name: methyl N-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]carbamate
Synonyms: methyl N-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]carbamate
Molecular Weight (g/mol): 278.12
Molecular Formula: C13H19BN2O4
InChi Key: QDYHEIDRTYYVQI-UHFFFAOYSA-N
SMILES: COC(=O)NC1=NC=C(C=C1)B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results