accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1073372-03-8

IUPAC Name: 4-[3-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]morpholine
Synonyms: 4-[3-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]morpholine
Molecular Weight (g/mol): 304.2
Molecular Formula: C16H25BN2O3
InChi Key: DGVAEMVUOIETDY-UHFFFAOYSA-N
SMILES: CC1=C(N=CC(=C1)B1OC(C)(C)C(C)(C)O1)N1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results