accessibility menu, dialog, popup

UserName

Viewing profile as user:
OO

CAS RN 1074-12-0

IUPAC Name: 2-oxo-2-phenylacetaldehyde
Synonyms: phenylglyoxal
Molecular Weight (g/mol): 134.13
Molecular Formula: C8H6O2
InChi Key: OJUGVDODNPJEEC-UHFFFAOYSA-N
SMILES: O=CC(=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results