accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1074-36-8

IUPAC Name: 4-sulfanylbenzoic acid
Synonyms: 2-mercaptobenzoic acid
Molecular Weight (g/mol): 154.18
Molecular Formula: C7H6O2S
InChi Key: LMJXSOYPAOSIPZ-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(S)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (1)
  • (4)
  • (1)
Quantity 
  • (1)
  • (2)
  • (3)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (2)
  • (3)
  • (3)
  • (1)
  • (4)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)