CAS RN 1074-43-7
IUPAC Name:
1-methyl-3-propylbenzene
Synonyms:
1-methyl-3-propylbenzene
Molecular Weight (g/mol):
134.22
Molecular Formula:
C10H14
InChi Key:
QUEBYVKXYIKVSO-UHFFFAOYSA-N
SMILES:
CCCC1=CC=CC(C)=C1
1
–
1
of
1
results
1
3-Propyltoluene 99.0+%, TCI America™
CAS: 1074-43-7 Molecular Formula: C10H14 Molecular Weight (g/mol): 134.22 MDL Number: MFCD00059247 InChI Key: QUEBYVKXYIKVSO-UHFFFAOYSA-N Synonym: 1-Methyl-3-propylbenzene PubChem CID: 14092 IUPAC Name: 1-methyl-3-propylbenzene SMILES: CCCC1=CC=CC(C)=C1