accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1074-51-7

IUPAC Name: N-cyclooctylidenehydroxylamine
Synonyms: N-cyclooctylidenehydroxylamine
Molecular Weight (g/mol): 141.21
Molecular Formula: C8H15NO
InChi Key: KTPUHSVFNHULJH-UHFFFAOYSA-N
SMILES: ON=C1CCCCCCC1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)