accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2COOOOCH2

CAS RN 107481-28-7

IUPAC Name: 9-(prop-2-enoyloxy)nonyl prop-2-enoate
Synonyms: 9-(prop-2-enoyloxy)nonyl prop-2-enoate
Molecular Weight (g/mol): 268.35
Molecular Formula: C15H24O4
InChi Key: PGDIJTMOHORACQ-UHFFFAOYSA-N
SMILES: C=CC(=O)OCCCCCCCCCOC(=O)C=C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results