accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHHN

CAS RN 1075-26-9

IUPAC Name: (1H-indol-6-yl)methanol
Synonyms: 1H-indol-6-ylmethanol
Molecular Weight (g/mol): 147.18
Molecular Formula: C9H9NO
InChi Key: WRMZOPANDOHWJU-UHFFFAOYSA-N
SMILES: OCC1=CC=C2C=CNC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results