accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1075-76-9

IUPAC Name: 3-(phenylamino)propanenitrile
Synonyms: 3-(phenylamino)propanenitrile
Molecular Weight (g/mol): 146.19
Molecular Formula: C9H10N2
InChi Key: FENJKTQEFUPECW-UHFFFAOYSA-N
SMILES: N#CCCNC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results