accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 107534-96-3

IUPAC Name: 1-(4-chlorophenyl)-4,4-dimethyl-3-[(1H-1,2,4-triazol-1-yl)methyl]pentan-3-ol
Synonyms: tebuconazole
Molecular Weight (g/mol): 307.82
Molecular Formula: C16H22ClN3O
InChi Key: PXMNMQRDXWABCY-UHFFFAOYNA-N
SMILES: CC(C)(C)C(O)(CCC1=CC=C(Cl)C=C1)CN1C=NC=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results