accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 107552-14-7

IUPAC Name: scandium(3+) trinitrate
Synonyms: scandium nitrate
Molecular Weight (g/mol): 230.97
Molecular Formula: N3O9Sc
InChi Key: DFCYEXJMCFQPPA-UHFFFAOYSA-N
SMILES: [Sc+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (11)
Grade 
  • (8)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (3)
  • (3)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (8)
  • (2)
  • (4)
  • (3)
  • (11)
  • (8)
  • (4)
  • (2)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)