accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1076-56-8

IUPAC Name: 2-methoxy-4-methyl-1-(propan-2-yl)benzene
Synonyms: 2-isopropyl-5-methylanisole
Molecular Weight (g/mol): 164.25
Molecular Formula: C11H16O
InChi Key: LSQXNMXDFRRDSJ-UHFFFAOYSA-N
SMILES: COC1=CC(C)=CC=C1C(C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results