accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOCH2

CAS RN 1076-96-6

IUPAC Name: methyl 4-ethenylbenzoate
Synonyms: methyl 4-ethenylbenzoate
Molecular Weight (g/mol): 162.19
Molecular Formula: C10H10O2
InChi Key: NUMHUJZXKZKUBN-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(C=C)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results