accessibility menu, dialog, popup

UserName

Viewing profile as user:
OI

CAS RN 107623-21-2

IUPAC Name: 1-(benzyloxy)-3-iodobenzene
Synonyms: 1-(benzyloxy)-3-iodobenzene
Molecular Weight (g/mol): 310.13
Molecular Formula: C13H11IO
InChi Key: QMKHOPJXDQAHBG-UHFFFAOYSA-N
SMILES: IC1=CC=CC(OCC2=CC=CC=C2)=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)