CAS RN 1077-01-6
IUPAC Name:
1-fluoro-3-(trifluoromethoxy)benzene
Synonyms:
1-fluoro-3-(trifluoromethoxy)benzene
Molecular Weight (g/mol):
180.1
Molecular Formula:
C7H4F4O
InChi Key:
AUKDFDQPJWJEDH-UHFFFAOYSA-N
SMILES:
FC1=CC=CC(OC(F)(F)F)=C1
1
–
1
of
1
results
1
1-Fluoro-3-(trifluoromethoxy)benzene 98.0+%, TCI America™
CAS: 1077-01-6 Molecular Formula: C7H4F4O Molecular Weight (g/mol): 180.102 MDL Number: MFCD00236323 InChI Key: AUKDFDQPJWJEDH-UHFFFAOYSA-N PubChem CID: 2777286 IUPAC Name: 1-fluoro-3-(trifluoromethoxy)benzene SMILES: C1=CC(=CC(=C1)F)OC(F)(F)F